Skip to Content
Merck
All Photos(1)

Documents

92220

Sigma-Aldrich

2′,4′,6′-Trimethylacetophenone

≥98.0% (GC)

Synonym(s):

2-Acetylmesitylene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3C6H2COCH3
CAS Number:
Molecular Weight:
162.23
Beilstein:
2044595
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98.0% (GC)

form

liquid

refractive index

n20/D 1.517 (lit.)
n20/D 1.517

bp

235-236 °C (lit.)
61-62 °C/0.1 mmHg (lit.)

density

0.975 g/mL at 25 °C (lit.)

SMILES string

CC(=O)c1c(C)cc(C)cc1C

InChI

1S/C11H14O/c1-7-5-8(2)11(10(4)12)9(3)6-7/h5-6H,1-4H3

InChI key

XWCIICLTKWRWCI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service