Skip to Content
Merck
All Photos(1)

Key Documents

805602

Sigma-Aldrich

3-(1-octyn-1-yl)-2-oxazolidinone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H17NO2
CAS Number:
Molecular Weight:
195.26
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

form

liquid

storage temp.

2-8°C

SMILES string

O=C1N(C#CCCCCCC)CCO1

InChI

1S/C11H17NO2/c1-2-3-4-5-6-7-8-12-9-10-14-11(12)13/h2-6,9-10H2,1H3

InChI key

BTHIUMSDMJKIBP-UHFFFAOYSA-N

Application

Nitrogen-substituted alkynes are strongly polarized alkynes and highly reactive building blocks which have been used for the design of a wide number of transformations. Precursor of highly reactive keteniminium ions and useful reagent for asymmetric synthesis.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Evano group focuses on natural product synthesis, copper catalysis, and hetero-substituted alkynes, emphasizing ynamides' synthetic potential and reactivity.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service