Skip to Content
Merck
All Photos(1)

Key Documents

746134

Sigma-Aldrich

Chitosan

high purity, Mv 40,000-60,000

Synonym(s):

KiOmedine-CsU®A, Deacetylated chitin, Poly(D-glucosamine)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352200

grade

high purity

mol wt

Mv 40,000-60,000

composition

Bacterial endotoxins, ≤10 EU/g
Beta-glucan, ≤5 % (w/w)
Chitosan content, ≥93 % (w/w)
Degree of acetylation, ≤40 mol %
Proteins, ≤1 % (w/w)

viscosity

5-200 mPa.s, 1 % in 1% acetic acid

solubility

acetic acid: 1% (solution)

storage temp.

2-8°C

InChI

1S/C56H103N9O39/c1-87-56(86)65-28-38(84)46(19(10-74)96-55(28)104-45-18(9-73)95-49(27(64)37(45)83)97-39-12(3-67)88-47(85)20(57)31(39)77)103-54-26(63)36(82)44(17(8-72)94-54)102-53-25(62)35(81)43(16(7-71)93-53)101-52-24(61)34(80)42(15(6-70)92-52)100-51-23(60)33(79)41(14(5-69)91-51)99-50-22(59)32(78)40(13(4-68)90-50)98-48-21(58)30(76)29(75)11(2-66)89-48/h11-55,66-85H,2-10,57-64H2,1H3,(H,65,86)/t11-,12-,13-,14-,15-,16-,17-,18-,19-,20-,21-,22-,23-,24-,25-,26-,27-,28-,29-,30-,31-,32-,33-,34-,35-,36-,37-,38-,39-,40-,41-,42-,43-,44-,45-,46-,47-,48+,49+,50+,51+,52+,53+,54+,55+/m1/s1

InChI key

FLASNYPZGWUPSU-SICDJOISSA-N

Looking for similar products? Visit Product Comparison Guide

General description

KiOmedine®-CsU is an ultra-pure chitosan of non-animal origin, produced from white mushrooms (Agaricus bisporus), in accordance with cGMP.
The crystalline structure is in the α-form. The linkage between glucosamine units is β-(1,4).

Application

These chitosans are high-value; medical grade polymers manufactured under cGMP conditions; commonly used in surface modification; tissue engineering; and drug delivery.
Forms gels with multivalent anions. Gives clear solutions that dry to strong, clear films.

Features and Benefits

Biocompatible, antibacterial and environmentally friendly polyelectrolyte with a variety of applications including water treatment, chromatography, additives for cosmetics, textile treatment for antimicrobial activity, novel fibers for textiles, photographic papers, biodegradable films, biomedical devices, and microcapsule implants for controlled release in drug delivery.
Biocompatible, antibacterial and environmentally friendly polyelectrolyte.

Legal Information

Product of KitoZyme
Kalignost is a registered trademark of Heyl Chemisch-Pharmazeutische Fabrik GmbH & Co. KG
KiOmedine is a registered trademark of KitoZyme, S.A.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liming Hu et al.
Nanoscale, 5(8), 3103-3111 (2013-03-22)
Within the past few years, chitosan-based drug delivery vehicles have become some of the most attractive to be studied. In contrast to all other polysaccharides, chitosan has demonstrated its unique characteristics for drug delivery platforms, including its active primary amino
Abdelkrim El Kadib et al.
Journal of nanoscience and nanotechnology, 14(1), 308-331 (2014-04-16)
Polysaccharides are a new class of pervasive biopolymers that display many advantages including wide availability, sustainability, inherent inclusion of chemical functionality, biocompatibility and biodegradability. Current efforts are focused on the catalytic transformation of these macromolecules into fuels and platform chemicals.
Hailing Liu et al.
Analytica chimica acta, 776, 24-30 (2013-04-23)
Based on the polyphosphonate-assisted coacervation of chitosan, a simple and versatile procedure for the encapsulation of proteins/enzymes in chitosan-carbon nanotubes (CNTs) composites matrix was developed. Scanning electron microscopy (SEM), transmission electron microscopy (TEM), X-ray photoelectron spectroscopy (XPS), energy dispersive spectrum
Milena Ignatova et al.
Macromolecular bioscience, 13(7), 860-872 (2013-06-12)
Chitosan is non-toxic, biocompatible, and biodegradable polysaccharide from renewable resources, known to have inherent antibacterial activity, which is mainly due to its polycationic nature. The combining of all assets of chitosan and its derivatives with the unique properties of electrospun
Alberto Busilacchi et al.
Carbohydrate polymers, 98(1), 665-676 (2013-08-31)
The idea of using chitosan as a functional delivery aid to support simultaneously PRP, stem cells and growth factors (GF) is associated with the intention to use morphogenic biomaterials to modulate the natural healing sequence in bone and other tissues.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service