Skip to Content
Merck
All Photos(1)

Key Documents

740144

Sigma-Aldrich

Mono(2-chloroallyl) oxalyl chloride

97%

Synonym(s):

2-Chloroallyl oxalochloride, Chlorooxoacetic acid 2-chloroallyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H4Cl2O3
CAS Number:
Molecular Weight:
182.99
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.466

density

1.371 g/mL at 25 °C

SMILES string

ClC(=C)COC(=O)C(Cl)=O

InChI

1S/C5H4Cl2O3/c1-3(6)2-10-5(9)4(7)8/h1-2H2

InChI key

DROVKFBGHINOBG-UHFFFAOYSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

199.9 °F

Flash Point(C)

93.3 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service