723959
6-Methyl-2-pyridinylboronic acid MIDA ester
Synonym(s):
2-(6-Methyl-2-pyridinyl)-6-methyl-1,3,6,2-dioxazaborocane-4,8-dione, 2-Methylpyridine-6-boronic acid MIDA ester, 6-Methylpyridine-2-boronic acid MIDA ester
About This Item
Recommended Products
form
solid
mp
264-268 °C
storage temp.
2-8°C
SMILES string
CN1CC(=O)OB(OC(=O)C1)c2cccc(C)n2
InChI
1S/C11H13BN2O4/c1-8-4-3-5-9(13-8)12-17-10(15)6-14(2)7-11(16)18-12/h3-5H,6-7H2,1-2H3
InChI key
AHYYPYSUORBPCY-UHFFFAOYSA-N
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service