Skip to Content
Merck
All Photos(1)

Key Documents

689564

Sigma-Aldrich

Potassium N-chloro-p-toluenesulfonamide

anhydrous, ≥98.0%

Synonym(s):

N-Chloro-4-methyl-benzenesulfonamide potassium salt, Chloramine T potassium salt, anhydrous

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H7ClKNO2S
CAS Number:
Molecular Weight:
243.75
MDL number:
UNSPSC Code:
12352000

Assay

≥98.0%
98.0-102.0% (RT)

quality

anhydrous

reaction suitability

reagent type: oxidant

loss

≤0.5% loss on drying

solubility

H2O: 50 mg/mL, clear, colorless to tan

SMILES string

Cc1ccc(cc1)S(=O)(=O)N(Cl)[K]

Application

Reactant for:
  • Carbonylation reactions

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service