688010
Poly[(1,4-divinylenephenylene)(2,4,6-triisopropylphenylborane)]
Synonym(s):
Boramer™-T01, Poly(1,4-divinylenephenylene tripylborane)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
mol wt
average Mn ~1379
mp
103-112 °C
fluorescence
λex 330 nm; λem 440 nm in chloroform
Application
n-type polymer semiconductor; HOMO = –6.65 eV, LUMO = –3.85 eV.
Legal Information
Manufactured by TDA research, Inc.
Boramer is a trademark of TDA Research, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Material Matters, 3, 11-11 (2007)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service