Skip to Content
Merck
All Photos(1)

Key Documents

649783

Sigma-Aldrich

Poly(3,4-ethylenedioxythiophene)-block-poly(ethylene glycol) solution

1 wt % dispersion in propylene carbonate, contains perchlorate as dopant

Synonym(s):

Aedotron C-PC, PEDOT-block-PEG, PEDOT-b-PEG

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352103

contains

perchlorate as dopant

concentration

1 wt % dispersion in propylene carbonate

conductivity

0.5-3 S/cm (bulk)

density

1.189 g/mL at 25 °C

Looking for similar products? Visit Product Comparison Guide

General description

Please follow this link for Aedotron/Oligotron selection guide table.

Application

Propylene carbonate can be used to cast films on many substrates, and is well suited for dipping and spray coating. It is a high boiling point solvent and may require heating (e.g. 80 °C) under vacuum to dry.
Conducting polymer. Useful as anti-static layers, hole-transport layers in OLED′s and various organic electronics applications (flexible circuits, capacitors, batteries).

Caution

Store this product at room temperature (do not refrigerate or freeze). Some settling will normally occur. Agitate, sonicate, and filter prior to use. A plug of glass wool in a pipette can be used for easy filtration. The shelf life of this product will vary depending on how it is stored and handled. Product should be discarded if heavy aggregation occurs, or if sonication / filtering are inadequate to produce a good casting solution.

Legal Information

Aedotron is a trademark of TDA Research, Inc.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

269.6 °F - closed cup

Flash Point(C)

132 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service