Skip to Content
Merck
All Photos(1)

Key Documents

573906

Sigma-Aldrich

Barium chromate

99.999% trace metals basis

Synonym(s):

Barium chromate (BaCrO4), Barium chromate(VI), Barium yellow, Lemon Yellow (inorganic pigment)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
BaCrO4
CAS Number:
Molecular Weight:
253.32
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Assay

99.999% trace metals basis

form

powder

impurities

≤20 ppm Trace metallic impurities

mp

210 °C (dec.) (lit.)

density

4.5 g/mL at 25 °C (lit.)

SMILES string

[Ba++].[O-][Cr]([O-])(=O)=O

InChI

1S/Ba.Cr.4O/q+2;;;;2*-1

InChI key

QFFVPLLCYGOFPU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 1B - Ox. Sol. 2

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sandra S Wise et al.
Environmental and molecular mutagenesis, 42(4), 274-278 (2003-12-16)
Hexavalent chromium (Cr(VI)) compounds are widely accepted as human lung carcinogens. However, there have been few investigations of the genotoxicity of Cr(VI) in human lung cells. Moreover, our knowledge of the effects of Cr(VI) in human lung cells is further
Sandra S Wise et al.
Environmental and molecular mutagenesis, 44(2), 156-162 (2004-07-28)
Particulate hexavalent chromium [Cr(VI)] compounds are well-established human lung carcinogens. However, their carcinogenic mechanisms are poorly understood as most investigators have used soluble Cr(VI) compounds. Recent work from our laboratory has found that barium chromate (BC) is also cytotoxic and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service