Skip to Content
Merck
All Photos(1)

Key Documents

572594

Sigma-Aldrich

Ammonium cerium(IV) nitrate on silica gel

Synonym(s):

Ceric ammonium nitrate on silica gel

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ce(NH4)2(NO3)6 · (SiO2)x
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.22

form

solid

Looking for similar products? Visit Product Comparison Guide

Application

Supported oxidizing reagent.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1 - Met. Corr. 1 - Ox. Sol. 2 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service