Skip to Content
Merck
All Photos(1)

Key Documents

567051

Sigma-Aldrich

Tetrakis(acetato)bis(cyclopentadienyl)divanadium(III)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C5H5V(OCCH3O)4VC5H5
CAS Number:
Molecular Weight:
468.25
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

form

solid

reaction suitability

core: vanadium
reagent type: catalyst

mp

200 °C (dec.) (lit.)

SMILES string

[CH]1[CH][CH][CH][CH]1.[CH]2[CH][CH][CH][CH]2.CC(=O)O[V]OC(C)=O.CC(=O)O[V]OC(C)=O

InChI

1S/2C5H5.4C2H4O2.2V/c2*1-2-4-5-3-1;4*1-2(3)4;;/h2*1-5H;4*1H3,(H,3,4);;/q;;;;;;2*+2/p-4

InChI key

GTNGKNSAYHMPTD-UHFFFAOYSA-J

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Promoter sequence and architecture determine expression variability and confer robustness to genetic variants.
Einarsson, et al.
eLife, 11 (2023)
James Budzak et al.
Nature communications, 13(1), 101-101 (2022-01-12)
A Variant Surface Glycoprotein (VSG) coat protects bloodstream form Trypanosoma brucei. Prodigious amounts of VSG mRNA (~7-10% total) are generated from a single RNA polymerase I (Pol I) transcribed VSG expression site (ES), necessitating extremely high levels of localised splicing.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service