Skip to Content
Merck
All Photos(1)

Key Documents

544361

Sigma-Aldrich

D-Carnitine

98%

Synonym(s):

(+)-β-Hydroxy-γ-(trimethylammonio)butyrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3N+CH2CH(OH)CH2COO-
CAS Number:
Molecular Weight:
161.20
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

98%

optical activity

[α]20/D +31°, c = 10 in H2O

mp

187-190 °C (dec.) (lit.)

SMILES string

C[N+](C)(C)C[C@@H](O)CC([O-])=O

Looking for similar products? Visit Product Comparison Guide

Application

Used as a building block for antidiabetic, microbicidal compounds.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P S Savle et al.
Bioorganic & medicinal chemistry letters, 9(17), 2545-2548 (1999-09-25)
Acylcarnitine analogues, (+)-6-Carboxylatomethyl-2-alkyl-4,4-dimethylmorpholinium (Z-n, where n = the number of carbons in the alkyl chain), synthesized in multi-gram quantities show in vitro activities as spermicides, anti-HIV agents, and inhibitors of the growth of Candida albicans. Activity improves with increasing chain

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service