440655
Boc-Asp(OBzl)-OH
98%
Synonym(s):
N-(tert-Butoxycarbonyl)-L-aspartic acid 4-benzyl ester, Boc-L-aspartic acid 4-benzyl ester
About This Item
Recommended Products
Assay
98%
optical activity
[α]20/D −19.5°, c = 2 in DMF
mp
98-102 °C (lit.)
SMILES string
[H][C@@](CC(=O)OCc1ccccc1)(NC(=O)OC(C)(C)C)C(O)=O
InChI
1S/C16H21NO6/c1-16(2,3)23-15(21)17-12(14(19)20)9-13(18)22-10-11-7-5-4-6-8-11/h4-8,12H,9-10H2,1-3H3,(H,17,21)(H,19,20)/t12-/m0/s1
InChI key
SOHLZANWVLCPHK-LBPRGKRZSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service