Skip to Content
Merck
All Photos(1)

Key Documents

429244

Sigma-Aldrich

Nylon 6/66

pellets

Synonym(s):

Poly(hexamethylene adipamide-co-caprolactam)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C18H37N3O5)n
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
Pricing and availability is not currently available.

abrasion resistance

5 mg (Taber, CS-17 wheel, 1kg, 50% RH, 1,000 cycles)

form

pellets

contains

heat stabilizer, nucleating agent and lubricant

hardness

100 (Rockwell R, ASTM D 785, 50% RH)
122 (Rockwell R, ASTM D 785, DAM)

mp

250-260 °C (lit.)

transition temp

Tm 250 °C (ASTM D 2117)

solubility

strong proton donor solvents, acetic acid and phenols: soluble

density

1.3 g/mL at 25 °C (lit.)

SMILES string

NCCCCCCN.O=C1CCCCCN1.OC(=O)CCCCC(O)=O

InChI

1S/C6H16N2.C6H11NO.C6H10O4/c7-5-3-1-2-4-6-8;8-6-4-2-1-3-5-7-6;7-5(8)3-1-2-4-6(9)10/h1-8H2;1-5H2,(H,7,8);1-4H2,(H,7,8)(H,9,10)

InChI key

TZYHIGCKINZLPD-UHFFFAOYSA-N

Application

General purpose thermoplastic polyamide.
Fibers for textiles, brushes and sutures cable sheathing and tubing and bearings, cams, gears and casings for tools and appliances.

Features and Benefits

Stiff and strong over a wide range of temperatures, excellent toughness and chemical and abrasion resistance. Attacked by strong acids and strong bases at elevated temperatures. Oxidized by chlorine and hydrogen peroxide.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service