Skip to Content
Merck
All Photos(1)

Key Documents

421464

Sigma-Aldrich

Zonyl® FSP fluorosurfactant

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002

refractive index

n20/D 1.358 (lit.)

bp

84 °C (lit.)

density

1.15 g/mL at 25 °C (lit.)

Legal Information

DuPont Product
Zonyl is a registered trademark of E. I. du Pont de Nemours and Company

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

75.2 °F - closed cup

Flash Point(C)

24 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chao Lu et al.
Analytical chemistry, 79(2), 666-672 (2007-01-16)
Gold nanoparticles (GNPs) capped with nonionic fluorosurfactant molecules (Zonyl FSN) were synthesized, and with the colloidal solution as a probe reagent, a new postcolumn colorimetric detection method for HPLC assay of homocysteine (Hcy) has been developed. The FSN-capped GNPs exhibited
Natalia Quinete et al.
Archives of environmental contamination and toxicology, 59(1), 20-30 (2010-01-08)
Perfluorooctanoic acid (PFOA) and perfluorooctane sulfonate (PFOS) are manmade, stable perfluorosurfactants. The properties of perfluoroalkylated compounds that cause them to persist in the environment are also the properties that made them attractive compounds for industrial usage for over 50 years.
Effects of laundering upon the removal of atrazine and metolachlor from cotton, cotton/polyester, and polyester fabrics treated with fluorochemical finishes.
A Shaw et al.
Bulletin of environmental contamination and toxicology, 66(3), 319-325 (2001-02-15)
V Caslavska et al.
Caries research, 25(1), 27-33 (1991-01-01)
After in vitro treatments of bovine enamel with NaF/Zonyl FSC and NH4F in-depth profiles of the chemical composition of the outermost zone of surface enamel were obtained with electron spectroscopy. The most important finding of the study was that after
Ingrid Machado de Andrade et al.
The International journal of prosthodontics, 25(2), 157-159 (2012-03-01)
The aim of this study was to evaluate the efficacy of experimental toothpastes for removing denture biofilm by means of a randomized crossover trial. Thirty volunteers brushed their dentures using a brush and four pastes: (1) Corega refreshing mint (control)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service