Skip to Content
Merck
All Photos(1)

Documents

40318

Sigma-Aldrich

Hydrogen peroxide solution

semiconductor grade ULSI PURANAL (Honeywell 17024), ≥30%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
H2O2
CAS Number:
Molecular Weight:
34.01
Beilstein:
3587191
MDL number:
UNSPSC Code:
12352304
PubChem Substance ID:

grade

semiconductor grade ULSI PURANAL (Honeywell 17024)

vapor pressure

23.3 mmHg ( 30 °C)

Assay

≥30%

CofA

specification on request

reaction suitability

reagent type: oxidant

storage temp.

2-8°C

SMILES string

OO

InChI

1S/H2O2/c1-2/h1-2H

InChI key

MHAJPDPJQMAIIY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

PURANAL is a trademark of Honeywell Specialty Chemicals Seelze GmbH

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stephen J D O'Keefe et al.
Nature communications, 6, 6342-6342 (2015-04-29)
Rates of colon cancer are much higher in African Americans (65:100,000) than in rural South Africans (<5:100,000). The higher rates are associated with higher animal protein and fat, and lower fibre consumption, higher colonic secondary bile acids, lower colonic short-chain
Shoichi Iriguchi et al.
Blood, 125(2), 370-382 (2014-10-29)
Although overexpression of T-bet, a master transcription factor in type-1 helper T lymphocytes, has been reported in several hematologic and immune diseases, its role in their pathogenesis is not fully understood. In the present study, we used transgenic model mice
Esther P M Tjin et al.
Cancer immunology research, 2(6), 538-546 (2014-06-05)
In this study, we investigated a large series of immune (escape) markers, relevant to T-cell function, as potential biomarkers for clinical outcome following immunotherapy. We retrospectively studied the expression of immune (escape) markers in metastatic melanoma tissues of 27 patients
Maheswari Jayakannan et al.
Journal of experimental botany, 66(7), 1865-1875 (2015-01-24)
The role of endogenous salicylic acid (SA) signalling cascades in plant responses to salt and oxidative stresses is unclear. Arabidopsis SA signalling mutants, namely npr1-5 (non-expresser of pathogenesis related gene1), which lacks NPR1-dependent SA signalling, and nudt7 (nudix hydrolase7), which
Osamu Sakai et al.
Investigative ophthalmology & visual science, 56(1), 538-543 (2015-01-13)
The purpose of the present study was to investigate the role of glutathione peroxidase 4 (GPx4) in conjunctival epithelial cells. An immortalized human conjunctival epithelial cell line was used. Cells were transfected with catalase, GPx1, GPx4, SOD1, SOD2, or control

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service