336866
Dichloromethylphosphine
technical grade, 90%
Synonym(s):
Methyldichlorophosphine
About This Item
Recommended Products
grade
technical grade
Assay
90%
reaction suitability
reagent type: ligand
refractive index
n20/D 1.496 (lit.)
bp
82 °C (lit.)
density
1.322 g/mL at 25 °C (lit.)
functional group
phosphine
SMILES string
CP(Cl)Cl
InChI
1S/CH3Cl2P/c1-4(2)3/h1H3
InChI key
CDPKWOKGVUHZFR-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Pyr. Liq. 1 - Skin Corr. 1B
Supplementary Hazards
Storage Class Code
4.2 - Pyrophoric and self-heating hazardous materials
WGK
WGK 3
Flash Point(F)
No data available
Flash Point(C)
No data available
Personal Protective Equipment
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service