Skip to Content
Merck
All Photos(1)

Key Documents

333727

Sigma-Aldrich

Bromomagnesium isopropylcyclohexylamide solution

1.0 M in THF

Synonym(s):

Bromo[N-(1-methylethyl)cyclohexanaminato]magnesium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H18BrMgN
CAS Number:
Molecular Weight:
244.45
MDL number:
UNSPSC Code:
12352103
Pricing and availability is not currently available.

concentration

1.0 M in THF

SMILES string

CC(C)N([Mg]Br)C1CCCCC1

Packaging

Packaged under nitrogen in Sure/Seal bottles

Legal Information

Sure/Seal is a trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

-2.2 °F - closed cup

Flash Point(C)

-19 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service