Skip to Content
Merck
All Photos(1)

Key Documents

299774

Sigma-Aldrich

1-Aminofluorene

99%

Synonym(s):

1-Fluorenamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H11N
CAS Number:
Molecular Weight:
181.23
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

mp

125-126 °C (lit.)

SMILES string

Nc1cccc-2c1Cc3ccccc-23

InChI

1S/C13H11N/c14-13-7-3-6-11-10-5-2-1-4-9(10)8-12(11)13/h1-7H,8,14H2

InChI key

CYSPWCARDHRYJX-UHFFFAOYSA-N

General description

1-Aminofluorene is pyrolysis product of 4-phenyl-oxindole.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The pyrolysis of oxindoles at 850? C.
Brown RFC and Butcher M.
Australian Journal of Chemistry, 25(1), 149-170 null
Tetyana Kobets et al.
Mutation research, 844, 10-24 (2019-07-22)
DNA-damaging activities of twenty-four structurally diverse unsubstituted and substituted cyclic compounds were assessed in embryo-fetal chicken livers. Formation of DNA adducts and strand breaks were measured using the nucleotide 32P-postlabelling (NPL) and comet assays, respectively. Unsubstituted monocyclic benzene, polycyclic fused

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service