Skip to Content
Merck
All Photos(1)

Key Documents

271659

Sigma-Aldrich

Histamine

96%

Synonym(s):

2-(4-Imidazolyl)ethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H9N3
CAS Number:
Molecular Weight:
111.15
Beilstein:
2012
EC Number:
MDL number:
UNSPSC Code:
12352100

Assay

96%

bp

167 °C/0.8 mmHg (lit.)

mp

83-84 °C (lit.)

SMILES string

NCCc1c[nH]cn1

InChI

1S/C5H9N3/c6-2-1-5-3-7-4-8-5/h3-4H,1-2,6H2,(H,7,8)

InChI key

NTYJJOPFIAHURM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Endogenous H1 and H2 histamine receptor agonist; H1 activation mobilizes Ca2+; H2 activation stimulates adenylate cyclase activity in neurons; activates nitric oxide synthetase; potent vasodilator.

replaced by

Product No.
Description
Pricing

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kwang-Youl Kim et al.
Journal of pharmaceutical and biomedical analysis, 172, 33-41 (2019-04-26)
The basophil histamine release test (HRT) is an important in vitro diagnostic assay to evaluate immunoglobin E (IgE)-mediated allergic responses. In this study, a bioanalytical LC-MS/MS method was developed and validated to quantify histamine in the leukocyte suspension from human
Amir Dekel et al.
Insect biochemistry and molecular biology, 110, 45-51 (2019-04-21)
The conservation of the mosquito indolergic receptors across the Culicinae and Anophelinae mosquito lineages, which spans 200 million years of evolution, is a testament to the central role of indolic compounds in the biology of these insects. Indole and skatole
Gui-Sheng Zhou et al.
Analytica chimica acta, 1107, 74-84 (2020-03-24)
A sensitive, rapid, precise and specific analytical method of hydrophilic interaction ultra-performance liquid chromatography coupled with triple-quadrupole linear ion-trap tandem mass spectrometry (HILIC-UHPLC-QTRAP®/MS2) combined with a high-efficiency and easy sample preparation technology of ultrasound-assisted ionic liquid dispersive liquid-liquid microextraction (UA-IL-DLLME)
Alessandra Misto et al.
Cell metabolism, 29(1), 91-102 (2018-10-16)
The conversion of lipolysis-derived fatty acids into ketone bodies (ketogenesis) is a crucial metabolic adaptation to prolonged periods of food scarcity. The process occurs primarily in liver mitochondria and is initiated by fatty-acid-mediated stimulation of the ligand-operated transcription factor, peroxisome

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service