Skip to Content
Merck
All Photos(1)

Key Documents

181108P

Avanti

Liver Extract Polar

Avanti Research - A Croda Brand

Synonym(s):

Liver Polar Lipid Extract (Bovine)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352211
NACRES:
NA.25

biological source

bovine (calf)

form

powder

packaging

pkg of 1 × 1 g (181108P-1g)
pkg of 1 × 100 mg (181108P-100mg)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

lipid extracts

shipped in

dry ice

storage temp.

−20°C

General description

Polar Lipid Extract is derived from the total lipid extract by precipitation with acetone followed by extraction of the acetone insoluble material with diethyl ether.

Application

Liver extract polar has been used to study the effects of mercury (Hg) on liver extract membrane fluidity.

Packaging

5 mL Clear Glass Sealed Ampule (181108P-100mg)
60 mL Amber Wide Mouth Screw Cap Glass Bottle (181108P-1g)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Inorganic mercury and cadmium induce rigidity in eukaryotic lipid extracts while mercury also ruptures red blood cells
Kerek E, et al.
Biochimica et Biophysica Acta - Biomembranes, 1860(3), 710-717 (2018)
Wenpeng Zhang et al.
Analytical chemistry, 92(9), 6719-6726 (2020-04-10)
Mapping the complete molecular composition of a lipidome is considered an important goal of lipidomics for unraveling pathways and mechanisms behind lipid homeostasis. Conventional dissociation methods of mass spectrometry (MS) usually cannot give detailed structural information on lipids such as
Wenbo Cao et al.
Nature communications, 11(1), 375-375 (2020-01-19)
Lipids play a pivotal role in biological processes and lipid analysis by mass spectrometry (MS) has significantly advanced lipidomic studies. While the structure specificity of lipid analysis proves to be critical for studying the biological functions of lipids, current mainstream

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service