Skip to Content
Merck
All Photos(4)

Key Documents

SAB4504698

Sigma-Aldrich

Anti-phospho-CRMP-2 (pThr509) antibody produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 62 kDa

species reactivity

mouse, human, rat

concentration

~1 mg/mL

technique(s)

ELISA: 1:1000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pThr509)

Gene Information

human ... DPYSL2(1808)

Immunogen

The antiserum was produced against synthesized peptide derived from human CRMP-2 around the phosphorylation site of Thr509.

Immunogen Range: 475-524

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Seigo Usuki et al.
Journal of oleo science, 67(1), 77-86 (2017-12-15)
The tuber of the konjac plant is a source enriched with GlcCer (kGlcCer), and has been used as a dietary supplement to improve the dry skin and itching that are caused by a deficiency of epidermal ceramide. Previously, we showed
Seigo Usuki et al.
Journal of oleo science, 67(1), 87-94 (2017-12-15)
Konjac ceramide (kCer) can be prepared by a chemoenzymatic method as previously published (Usuki, S.; Tamura, N.; Sakai, S.; Tamura, T.; Mukai, K.; Igarashi, Y. Biochem. Biophys. Rep. 5, 160-167 (2016)). Thus prepared kCer showed an activation effect on Sema3A

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service