- Konjac Ceramide (kCer) Regulates NGF-Induced Neurite Outgrowth via the Sema3A Signaling Pathway.
Konjac Ceramide (kCer) Regulates NGF-Induced Neurite Outgrowth via the Sema3A Signaling Pathway.
Journal of oleo science (2017-12-15)
Seigo Usuki, Noriko Tamura, Kohei Yuyama, Tomohiro Tamura, Katsuyuki Mukai, Yasuyuki Igarashi
PMID29238028
ABSTRACT
The tuber of the konjac plant is a source enriched with GlcCer (kGlcCer), and has been used as a dietary supplement to improve the dry skin and itching that are caused by a deficiency of epidermal ceramide. Previously, we showed chemoenzymatically prepared konjac ceramide has a neurite-outgrowth inhibitory effect that is very similar to that of Sema3A and is not seen with animal-type ceramides. While, it has been unclear whether kCer may act on Sema3A or TrkA signaling pathway. In the present study, we showed kCer induces phosphorylation of CRMP2 and microtubules depolymerization via Sema3A signaling pathway not TrkA. It is concluded that kCer may be a potential Sema3A-like agonist that activates Sema3A signaling pathway directly.
Sigma-Aldrich
Anti-CRMP2 antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-phospho-CRMP-2 (pThr509) antibody produced in rabbit, affinity isolated antibody