Williams' Medium E Formulation
Component | W1878 [1x] g/L | W4125 g/L | W4128 [1x] g/L |
---|---|---|---|
Inorganic Salts | |||
Calcium Chloride | 0.2 | 0.2 | 0.2 |
Cupric Sulfate • 5H2O | 0.0000001 | 0.0000001 | 0.0000001 |
Ferric Nitrate • 9H2O | 0.0000001 | 0.0000001 | 0.0000001 |
Magnesium Chloride • 4H2O | 0.0000001 | 0.0000001 | 0.0000001 |
Magnesium Sulfate (anhydrous) | 0.0977 | 0.0977 | 0.0977 |
Potassium Chloride | 0.4 | 0.4 | 0.4 |
Sodium Bicarbonate | 2.2 | — | 2.2 |
Sodium Chloride | 6.8 | 6.8 | 6.8 |
Sodium Phosphate Monobasic (anhydrous) | 0.122 | 0.122 | 0.122 |
Zinc Sulfate • 7H2O | 0.0000002 | 0.0000002 | 0.0000002 |
Amino Acids | |||
L-Alanine | 0.09 | 0.09 | 0.09 |
L-Arginine (free base) | 0.05 | 0.05 | 0.05 |
L-Asparagine • H2O | 0.02 | 0.02 | 0.02 |
L-Aspartic Acid | 0.03 | 0.03 | 0.03 |
L-Cysteine (free acid) | 0.04 | 0.04 | 0.04 |
L-Cystine | 0.02 | 0.02 | 0.02 |
L-Glutamic Acid | 0.0445 | 0.0445 | 0.0445 |
L-Glutamine | — | 0.292 | — |
Glycine | 0.05 | 0.05 | 0.05 |
L-Histidine (free base) | 0.015 | 0.015 | 0.015 |
L-Isoleucine | 0.05 | 0.05 | 0.05 |
L-Leucine | 0.075 | 0.075 | 0.075 |
L-Lysine • HCl | 0.08746 | 0.08746 | 0.08746 |
L-Methionine | 0.015 | 0.015 | 0.015 |
L-Phenylalanine | 0.025 | 0.025 | 0.025 |
L-Proline | 0.03 | 0.03 | 0.03 |
L-Serine | 0.01 | 0.01 | 0.01 |
L-Threonine | 0.04 | 0.04 | 0.04 |
L-Tryptophan | 0.01 | 0.01 | 0.01 |
L-Tyrosine • 2Na • 2H2O | 0.05045 | 0.05045 | 0.05045 |
L-Valine | 0.05 | 0.05 | 0.05 |
Vitamins | |||
Ascorbic Acid • Na | 0.00227 | 0.00227 | 0.00227 |
D-Biotin | 0.0005 | 0.0005 | 0.0005 |
Calciferol | 0.0001 | 0.0001 | 0.0001 |
Choline Chloride | 0.0015 | 0.0015 | 0.0015 |
Folic Acid | 0.001 | 0.001 | 0.001 |
myo-Inositol | 0.002 | 0.002 | 0.002 |
Menadione (sodium bisulfite) | 0.00001 | 0.00001 | 0.00001 |
Niacinamide | 0.001 | 0.001 | 0.001 |
D-Pantothenic Acid (hemicalcium) | 0.001 | 0.001 | 0.001 |
Pyridoxal • HCl | 0.001 | 0.001 | 0.001 |
Retinol Acetate | 0.0001 | 0.0001 | 0.0001 |
Riboflavin | 0.0001 | 0.0001 | 0.0001 |
Thiamine • HCl | 0.001 | 0.001 | 0.001 |
DL-α-Tocopherol Phosphate • Na | 0.00001 | 0.00001 | 0.00001 |
Vitamin B12 | 0.002 | 0.0002 | 0.0002 |
Other | |||
D-Glucose | 2 | 2 | 2 |
Glutathione (reduced) | 0.00005 | 0.00005 | 0.00005 |
Methyl Linoleate | 0.00003 | 0.00003 | 0.00003 |
Phenol Red • Na | — | 0.0107 | 0.0107 |
Pyruvic Acid • Na | 0.025 | 0.025 | 0.025 |
Add | |||
L-Glutamine | 0.292 | — | 0.292 |
Sodium Bicarbonate | — | 2.2 | — |