LC/MS Analysis of Nucleotides SeQuant® ZIC®-cHILIC – Effect of pH
Figure 1.
Conditions | |
---|---|
Instrument | Agilent® 6530 Q-TOF LC/MS |
Column | SeQuant® ZIC®-cHILIC, 10 cm x 2.1 mm, 3.0 µm |
Mobile Phase | [A] acetonitrile; [B] 50 mM ammonium acetate, pH as indicated with acetic acid in water (50 mM ammonium acetate, pH 6.93 in water not adjusted) |
Gradient | 26 to 27% B in 10 min; to 27 to 35% B in 10 min; to 26% B in 0.1 min; hold at 26% B for 9.9 min |
Flow Rate | 0.3 mL/min |
Pressure | 986 psi |
Column Temp. | 40 °C |
Detector | Q-TOF (ESI-negative) |
Detector Other | XIC of the following m/z values: AMP-346, ADP-426, ATP-506, CMP-322, CDP-402, CTP-482, GMP-362, GDP-442, GTP-522, UMP-323, UDP-403, UTP-483 |
Injection | 10 µL |
Description | |
---|---|
Analysis Note | ZIC®-cHILIC is a densely bonded zwitterionic stationary phase with phosphorylcholine functional groups covalently attached to silica. The stationary phase has an overall 1:1 charge balance between the permanent negative phosphate groups close to the silica support and the distal positively quaternary ammonium groups. It has though a slight positive surface charge because of the spatial arrangement. A true zwitterionic stationary phase, with permanent strong acidic and basic functionalities, provide hydrophilic partitioning combined with weak ionic interactions and makes the phase virtually pH independent as shown in this analysis. |
Featured Industry | Life Science and Biopharma |
Legal Information | SeQuant is a trademark of Merck KGaA, Darmstadt, Germany LiChrosolv is a trademark of Merck KGaA, Darmstadt, Germany LiChropur is a trademark of Merck KGaA, Darmstadt, Germany |
Application No. | ASC140218I |
Materials
申し訳ございませんが、想定外のエラーが発生しました。
Network error: Failed to fetch