HPLC Analysis of GMP, GDP and GTP on SeQuant ZIC-HILIC
Conditions | |
---|---|
column | SeQuant ZIC-HILIC, PEEK, 100 x 4.6 mm, 5 µm, 200 Ã… (1.50453.0001) |
mobile phase | [A] acetonitrile; [B] ammonium acetate; (70:30, A,B) |
flow rate | 0.7 mL/min |
injection | 5 µL |
detector | 259 nm |
Description | |
---|---|
Categories | Analytical Chromatography, Nucleotides |
Featured Industry | Life Science and Biopharma |
suitability | application for HPLC |
Materials
申し訳ございませんが、想定外のエラーが発生しました。
Network error: Failed to fetch