HomeLarge Molecule HPLCHPLC Analysis of LMW Heparin-Dalteparin and Enoxaparin Analysis on Zenix®-100
HPLC Analysis of LMW Heparin-Dalteparin and Enoxaparin Analysis on Zenix®-100
Conditions | |
---|---|
Column | Zenix SEC-100, 30 cm x 7.8 mm I.D., 3 µm particles, 100 Ã… (Z777008) |
Column temp. | 35 °C |
Mobile phase | Sodium sulfate, 2.84%, pH 5.0 |
Flow rate | 0.5 mL/min |
Sample | 10 mg/mL Dalteparin in water, pH 6.0, MW 3,000 - 8,000 Da (1st chromatogram) |
Sample | 10 mg/mL Enoxaparin in mobile phase, MW 3,000 - 8,000 Da (2nd chroatogram) |
Injection | 25 µL |
Detector | UV 234 nm and RI |
Description | |
---|---|
Analysis Note | High resolution separations of low molecular weight heparin-daletparin and enoxaparin can be achived using Zenix SEC-100 columns. |
Categories | Analytical Chromatography, Anticoagulants |
Featured Industry | Life Science and Biopharma |
Legal Information | Zenix is a registered trademark of Sepax Technologies |
Suitability | Application for HPLC |
Materials
申し訳ございませんが、想定外のエラーが発生しました。
Network error: Failed to fetch