コンテンツへスキップ
Merck
  • Simultaneous detection of pH value and glucose concentrations for wound monitoring applications.
Simultaneous detection of pH value and glucose concentrations for wound monitoring applications.
Biosensors & bioelectronics (2016-08-31)
D A Jankowska, M B Bannwarth, C Schulenburg, G Faccio, K Maniura-Weber, R M Rossi, L Scherer, M Richter, L F Boesel
要旨
Aging population and longer life expectancy are the main reasons for an increasing number of patients with wound problems. Although the interest in wound care increases continuously, wound management still remains a challenge mainly due to the higher occurrence of chronic wounds, which require intensive care and constant monitoring. Here, we demonstrate a fluorescent sensing system to monitor the wound status and to distinguish between an autonomously healing and a chronic wound at an early stage. The system allows monitoring two of the most relevant fluctuating wound parameters during the healing process which are pH and glucose concentration. A fluorescent pH indicator dye, carboxynaphthofluorescein, and a metabolite-sensing enzymatic system, based on glucose oxidase and horseradish peroxidase, were immobilized on a biocompatible polysaccharide matrix to develop a functional hydrogel coating for wound monitoring. The changes in metabolite and enzyme concentration in artificial wound extract were converted into a fluorescent signal.
材料
製品番号
ブランド
製品内容
Sigma-Aldrich
5(6)-Carboxynaphthofluorescein, BioReagent, suitable for fluorescence, ≥90% (HPLC)
現在、価格および在庫状況を閲覧できません。