テクニック
UHPLC: suitable
試験パラメータ
column: BIOshell A160 Peptide C18, 15 cm x 2.1 mm I.D., 2.7 μm particles (66905-U)
mobile phase: [A] 75:25 (8 mM ammonium formate, pH 2.6 with formic acid:acetonitrile); [B] 50:50 (8 mM ammonium formate, pH 2.6 with formic acid:acetonitrile); (77:23, A:B)
flow rate: 0.2 mL/min
column temp.: 75 °C
detector: MS, ESI(+), TIC 100-3000 m/z
injection: 0.5 μL
sample: Mixture of six insulin variants, 100 μg/mL, 8 mM ammonium formate, pH 2.6 with formic acid
適合性
application for UHPLC
アプリケーション
life science and biopharma
アナリシスノート
BIOshell A160 Peptide C18 is an ideal column for reversed-phase separations of small proteins to large peptide sequences. There is over 75% sequence similarity between all six of the insulin variants in the sample; therefore, separation of these variants is achieved through the subtle differences in the polarities of surface residues on the peptides. The use of ammonium formate as a mobile phase modifier yields more efficient peaks and less signal suppression during electrospray ionization (ESI) than some traditional ion-pair reagents (trifluoroacetic acid, formic acid).
法的情報
アナライト
詳細
Insulin-Glargine
Insulin-Bovine
Insulin-Asp
Insulin-Lispro
Insulin-Human
Insulin-Porcine
最新バージョンのいずれかを選択してください:
ライフサイエンス、有機合成、材料科学、クロマトグラフィー、分析など、あらゆる分野の研究に経験のあるメンバーがおります。.
製品に関するお問い合わせはこちら(テクニカルサービス)