おすすめの製品
アッセイ
≥98% (TLC)
mp
197-200 °C (dec.)
保管温度
−20°C
SMILES記法
Cl.Cl.NCC[C@H](N)C(O)=O
InChI
1S/C4H10N2O2.2ClH/c5-2-1-3(6)4(7)8;;/h3H,1-2,5-6H2,(H,7,8);2*1H/t3-;;/m0../s1
InChI Key
CKAAWCHIBBNLOJ-QTNFYWBSSA-N
類似した製品をお探しですか? 訪問 製品比較ガイド
以下による置換
シグナルワード
Danger
危険有害性情報
危険有害性の分類
Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3
保管分類コード
13 - Non Combustible Solids
WGK
WGK 3
引火点(°F)
Not applicable
引火点(℃)
Not applicable
個人用保護具 (PPE)
dust mask type N95 (US), Eyeshields, Gloves
Paul Alan Cox et al.
Amyotrophic lateral sclerosis : official publication of the World Federation of Neurology Research Group on Motor Neuron Diseases, 10 Suppl 2, 109-117 (2009-12-16)
Veterans of the 1990-1991 Gulf War have been reported to have an increased incidence of amyotrophic lateral sclerosis (ALS) compared to personnel who were not deployed. An excess of ALS cases was diagnosed in Gulf War veterans younger than 45
Discovery of α,β- and α,γ-diamino acid scaffolds for the inhibition of M1 family aminopeptidases.
Rajesh Gumpena et al.
ChemMedChem, 6(11), 1971-1976 (2011-10-26)
Hang-Yeon Weon et al.
Journal of microbiology (Seoul, Korea), 50(1), 161-165 (2012-03-01)
The bacterial strain M1T8B10(T) was isolated from cow dung in Suwon, Republic of Korea. The strain was a Gram stain-positive rod, nonmotile, and non-spore-forming. According to 16S rRNA gene sequence analysis, the strain fell within the clade of the genus
Undine Behrendt et al.
International journal of systematic and evolutionary microbiology, 58(Pt 11), 2574-2578 (2008-11-06)
In the course of studying the composition of the microbial community in the phyllosphere of potato plants, a Gram-positive bacterium, strain K 70/01(T), was isolated. On the basis of 16S rRNA gene sequence analysis, it was shown that the investigated
G S N Reddy et al.
International journal of systematic and evolutionary microbiology, 58(Pt 9), 2229-2234 (2008-09-05)
Strain PON10(T) is a yellow-pigmented, Gram-positive, aerobic, motile, rod-shaped bacterium isolated from the Pindari glacier of the Indian Himalayas. The cell-wall peptidoglycan contained DL-diaminobutyric acid as the diamino acid. The predominant fatty acids were anteiso-C(15:0), anteiso-C(17:0) and iso-C(16:0) and the
ライフサイエンス、有機合成、材料科学、クロマトグラフィー、分析など、あらゆる分野の研究に経験のあるメンバーがおります。.
製品に関するお問い合わせはこちら(テクニカルサービス)