おすすめの製品
SMILES記法
CC(O)CC(O)=O
InChI
1S/C12H20O6/c1-8(14)6-11(15)18-10(3)7-12(16)17-9(2)4-5-13/h5,8-10,14H,4,6-7H2,1-3H3
InChI Key
QLACRIKFZRFWRU-UHFFFAOYSA-N
類似した製品をお探しですか? 訪問 製品比較ガイド
その他情報
光学的に純粋な(R)-3-ヒドロキシ酪酸とそのメチルエステルや(R)-1,3-ブタンジオ-ルの合成に用いられる有用な出発物質です。
以下による置換
最新バージョンのいずれかを選択してください:
D. Seebach et al.
Helvetica Chimica Acta, 65, 495-495 (1982)
Geeta Gahlawat et al.
Applied biochemistry and biotechnology, 169(3), 821-831 (2013-01-01)
Culture fluorescence measurement is an indirect and non-invasive method of biomass estimation to assess the metabolic state of the microorganism in a fermentation process. In the present investigation, NAD(P)H fluorescence has been used for on-line in situ characterization of metabolic
Lauro Xavier Filho et al.
Journal of nanoscience and nanotechnology, 13(7), 4715-4719 (2013-08-02)
Nanotholits is an osteoinductor or be, stimulates the bone regeneration, enabling bigger migration of the cells for formation of the bone tissue regeneration mainly because nanotholits are rich in minerals considered essential to the bone mineralization process on a protein
D. Seebach et al.
Organic Syntheses, 71, 39-39 (1993)
Klaus Heuner et al.
Methods in molecular biology (Clifton, N.J.), 954, 163-181 (2012-11-15)
Metabolic pathways and fluxes can be analyzed under in vivo conditions by incorporation experiments using general (13)C-labeled precursors. On the basis of the isotopologue compositions in amino acids or other metabolites, the incorporation rates of the supplied precursors and the
ライフサイエンス、有機合成、材料科学、クロマトグラフィー、分析など、あらゆる分野の研究に経験のあるメンバーがおります。.
製品に関するお問い合わせはこちら(テクニカルサービス)