表示を変更する
| あなたへ/カタログ番号 | 在庫状況 | 単価 |
|---|---|---|
1 ea | カートの在庫状況を確認する | ¥40,400 |
この商品について
NACRES:
NB.51
UNSPSC Code:
41100000
Joint:
joint: ST/NS24/40
Packaging:
pkg of 1 ea
¥40,400
カートの在庫状況を確認する
joint
joint: ST/NS24/40
packaging
pkg of 1 ea
manufacturer/tradename
Synthware P140100M
capacity
100 mL , 100 mL
fitting
GL 25
General description
A connection of 8 mm O.D. is sealed on the inner joint for connection to an inner gas. With a fritted disc of coarse or medium porosity to support resin. Top of the vessel has GL thread and a PTFE lined screw cap. PTFE stopcock has a 2 mm bore with the arm being 8 mm O.D.
Legal Information
Synthware is a trademark of Kemtech America Inc.
1 of 1
当該品目 | |||
|---|---|---|---|
| capacity 100 mL , 100 mL | capacity 10 mL , 10 mL | capacity 50 mL , 50 mL | capacity 100 mL , 100 mL |
| manufacturer/tradename Synthware P140100M | manufacturer/tradename Synthware P140010M | manufacturer/tradename Synthware P140050M | manufacturer/tradename Synthware P130100C |
| joint joint: ST/NS24/40 | joint joint: ST/NS24/40 | joint joint: ST/NS24/40 | joint - |
| packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea |
| fitting GL 25 | fitting GL 14 | fitting GL 25 | fitting GL 25 |
Still not finding the right product?
Explore all of our products under Synthware™ peptide synthesis vessel with fritted disc, T-bore PTFE stopcock and vacuum take-off