Sigma-Aldrich
Methyl methacrylate, contains ≤30 ppm MEHQ as inhibitor, 99%
Pricing and availability is not currently available.
Sigma-Aldrich
Tetrahydrofuran, JIS special grade, ≥99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Tetrahydrofuran, suitable for HPLC, contains no stabilizer
Pricing and availability is not currently available.
Sigma-Aldrich
Tetrahydrofuran, anhydrous, contains 250 ppm BHT as inhibitor, ≥99.9%
Pricing and availability is not currently available.
Sigma-Aldrich
2-Hydroxyethyl methacrylate, ≥99%, contains ≤50 ppm monomethyl ether hydroquinone as inhibitor
Pricing and availability is not currently available.
Sigma-Aldrich
Copper(I) chloride, ≥99.995% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Methyl methacrylate, SAJ first grade, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Tetrahydrofuran, SAJ first grade, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
2,2′-Dipyridyl, JIS special grade, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Copper(I) chloride, JIS special grade, ≥95.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Diisopropyl ether, JIS special grade, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Diisopropyl ether, SAJ first grade, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
2-Hydroxyethyl methacrylate, embedding medium (for microscopy)
Pricing and availability is not currently available.
Sigma-Aldrich
Tetrahydrofuran, anhydrous, ≥99.9%, inhibitor-free
Pricing and availability is not currently available.
Sigma-Aldrich
Aluminum oxide, mesoporous, MSU-X (wormhole), average pore size 3.8 nm
Pricing and availability is not currently available.
Sigma-Aldrich
2-Hydroxyethyl methacrylate, contains ≤250 ppm monomethyl ether hydroquinone as inhibitor, 97%
Pricing and availability is not currently available.
Sigma-Aldrich
Tetrahydrofuran, suitable for HPLC, ≥99.9%, inhibitor-free
Pricing and availability is not currently available.
Sigma-Aldrich
Copper(I) chloride, AnhydroBeads™, ≥99.99% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, suitable for HPLC
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Diisopropyl ether, anhydrous, 99%, contains either BHT or hydroquinone as stabilizer
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, for residue analysis
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, SAJ special grade
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, NMR reference standard
Pricing and availability is not currently available.
Sigma-Aldrich
Aluminum oxide, 99.997% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
2,2′-Bipyridyl, ReagentPlus®, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
Aluminum oxide, nanopowder, 13 nm primary particle size (TEM), 99.8% trace metals basis
Pricing and availability is not currently available.