Merck
  • Regulated bioanalysis of conformers - A case study with ASP2151 in dog plasma and urine.
Regulated bioanalysis of conformers - A case study with ASP2151 in dog plasma and urine.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2015-06-21)
Yoshiaki Ohtsu, Shohei Otsuka, Takeshi Nakamura, Kiyoshi Noguchi
ABSTRACT
We developed and validated bioanalytical methods for a potent helicase-primase inhibitor ASP2151 that has two conformers. These conformers elute as unseparated broad peaks under ordinary high-performance liquid chromatographic conditions, indicating discernable differences in hydrophobicity. We observed that column temperature and mobile phase pH have no effect on these peaks and that conformers form a single symmetrical peak when tetrahydrofuran is added to the mobile phase. In addition, we needed to develop semi-automated methods where inter-conversion of the conformers is unlikely to cause sample-to-sample extraction variability. Briefly, following the addition of deuterium-labeled ASP2151 as an internal standard (IS), dog plasma samples or acetonitrile-added urine samples were filtrated. The filtrates were then injected into a column-switching liquid chromatography-tandem mass spectrometry (LC-MS/MS) system and trapped onto an extraction column. Extracts were back-flushed onto an analytical C18 column (4.6×50mm, 3μm) with a mobile phase consisting of methanol, tetrahydrofuran, and 20mmol/L ammonium acetate (45:5:50, v/v/v). The eluent was monitored in the negative atmospheric pressure chemical ionization mode. The calibration curve was linear over a range of 5-1000ng/mL for plasma and 0.5-100μg/mL for urine. Validation data met the acceptance criteria in accordance with regulatory guidance and demonstrated that these methods were selective, accurate, and reproducible. In addition, the present methods were successfully applied to a pharmacokinetic study in dogs.
Sigma-Aldrich
Tetrahydrofuran, JIS special grade, ≥99.5%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Tetrahydrofuran, suitable for HPLC, contains no stabilizer
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Tetrahydrofuran, anhydrous, contains 250 ppm BHT as inhibitor, ≥99.9%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Tetrahydrofuran, SAJ first grade, ≥99.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ammonium acetate, SAJ first grade, ≥97.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ammonium acetate, JIS special grade, ≥97.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ammonium acetate solution, 50 % (w/v), suitable for iron determination
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ammonium acetate solution, 50 % (w/v), for copper determination
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ammonium acetate solution, for molecular biology, 7.5 M
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Tetrahydrofuran, anhydrous, ≥99.9%, inhibitor-free
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Tetrahydrofuran, suitable for HPLC, ≥99.9%, inhibitor-free
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, suitable for HPLC
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, anhydrous, 99.8%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ammonium acetate, for molecular biology, ≥98%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, for residue analysis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, SAJ special grade
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, NMR reference standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetonitrile, for residue analysis, JIS 5000