Custom Rat / Mouse Monoclonal Antibody Production
Produced by obtaining antibody production cells with the Iliac lymph node method. The method can obtain a large number of positive clones with a shorter immunization period than the conventional spleen method.
Our service using the iliac lymph node method allows positive clones to be established in 1.5 months (approximately 45 days) at the earliest. The method can also obtain a greater number of positive clones than the spleen method, thus meeting customer needs in a variety of applications, including ELISA, immunostaining, flow cytometry, neutralization, and Western blotting.
Positive clones can be established in 1.5 months at the earliest
By using the iliac lymph node method, we can minimize the number of immunizations compared with that required for the conventional method. This greatly shortens the production period and enables the rapid delivery of monoclonal antibodies.
Customer-initiated work testing of antibodies
We will select positive clones in the process, and then provide the culture supernatant to the customer for evaluation of whether the antibody actually works in the customer's application.
Also, the entire process will be divided into 3 steps, and for each step, the customer will decide whether or not to proceed to the next step. Billing will occur at the end of each step.
Purified antibodies and hybridomas will be delivered / Intellectual property rights will be completely transferred
After delivery, mass cultivation and purification by the customer are also possible.
It is thus up to the customer to further expand the range of applications, such as product commercialization, and distribution and transfer to co-researchers.
Flow from ordering to delivery
Based on information obtained from the inquiries, a dedicated antibody expert will have a technical meeting with the customer to prepare the desired antibody.
A work schedule for the production of the antibody will be prepared and shown with the fee, and the customer can place the order if satisfied. A full technical support system will be in place during the process to provide progress reports and address requests for additional works.
1. Contact
*Account registration is required to place an order. Register here.
2. Primary hearing
A technical service person will explain about the antibody preparation service and confirm your request regarding the desired antibody.
3. Final meeting
Engineer visit and process setup.
Step 1
- Antigen synthesis and immunization of rats/mice
- Hybridoma establishment and seeding in 96-well plates
- 1st screening
Deliverables
- Work report (ELISA results), culture supernatant from positive wells
Step 2
- Cloning from customer designated wells
- 2nd screening
Deliverables
- Work report (ELISA results), culture supernatant from positive clones
Step 3
- Culturing of customer designated hybridomas
- Delivery of cultured cells
Deliverables
- Work report (entire work), cultured hybridomas
COMPARISON OF ILIAC LYMPH NODE METHOD WITH CONVENTIONAL METHOD | ||
---|---|---|
Iliac Lymph Node Method | Conventional Method | |
Number of immunizations | Once (~ 400 μg) | Several times |
Lymphocytes | Iliac nodes | Mouse spleen |
Number of lymphocytes | 1x108to3x108 cells | 1x108 cells |
Positive clone frequency | + + + + + | + |
From immunization to cell fusion | 2-3 weeks | 2 months |
Time until monoclonal antibody is esablished | 1 to 1.5 months | 3 to 6 months |
Related Product Resources
Article: Custom monoclonal antibody production: Frequently asked questions (FAQs)
Customer Service