90453
3,3′-Thiodipropionic acid
analytical standard
Synonym(s):
Bis(2-carboxyethyl) sulfide
About This Item
Recommended Products
grade
analytical standard
Assay
≥96.5% (GC)
form
powder
shelf life
limited shelf life, expiry date on the label
mp
131-134 °C (lit.)
SMILES string
OC(=O)CCSCCC(O)=O
InChI
1S/C6H10O4S/c7-5(8)1-3-11-4-2-6(9)10/h1-4H2,(H,7,8)(H,9,10)
InChI key
ODJQKYXPKWQWNK-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Recommended products
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
262.4 °F - closed cup
Flash Point(C)
128 °C - closed cup
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
ISHL Indicated Name
Substances Subject to be Indicated Names
ISHL Notified Names
Substances Subject to be Notified Names
JAN Code
90453-VAR:
90453-BULK:
90453-100MG:
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service