![3,3′-Thiodipropionic acid analytical standard](/deepweb/assets/sigmaaldrich/product/structures/320/898/fcd5f40d-c698-41a8-9b59-1e9187f4b96d/640/fcd5f40d-c698-41a8-9b59-1e9187f4b96d.png)
3,3′-Thiodipropionic acid
![3,3′-Thiodipropionic acid analytical standard](/deepweb/assets/sigmaaldrich/product/structures/320/898/fcd5f40d-c698-41a8-9b59-1e9187f4b96d/640/fcd5f40d-c698-41a8-9b59-1e9187f4b96d.png)
Synonym(s):
Bis(2-carboxyethyl) sulfide, 3,3′-Thiodipropionic acid
Linear Formula:
S(CH2CH2COOH)2
CAS Number:
Molecular Weight:
178.21
Products
Shipping
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing