Skip to Content
Merck
All Photos(2)

Key Documents

Safety Information

T2763

Sigma-Aldrich

Thymol iodide

Powder

Synonym(s):

4,4′-Bis(iodooxy)-2,2′-dimethyl-5,5′-bis(1-methylethyl)-1,1′-biphenyl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H24I2O2
CAS Number:
Molecular Weight:
550.21
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

product name

Thymol iodide,

form

powder

Quality Level

εmax

≥320 at 241-246 nm in chloroform

antibiotic activity spectrum

fungi

application(s)

diagnostic assay manufacturing
hematology
histology

Mode of action

DNA synthesis | interferes

storage temp.

2-8°C

SMILES string

CC(C)c1cc(c(C)cc1OI)-c2cc(C(C)C)c(OI)cc2C

InChI

1S/C20H24I2O2/c1-11(2)15-9-17(13(5)7-19(15)23-21)18-10-16(12(3)4)20(24-22)8-14(18)6/h7-12H,1-6H3

InChI key

SHOKWSLXDAIZPP-UHFFFAOYSA-N

Biochem/physiol Actions

Iodinated thymol with mild antiseptic activity

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

ISHL Indicated Name

Substances Subject to be Indicated Names

ISHL Notified Names

Substances Subject to be Notified Names

JAN Code

T2763-VAR:
T2763-BULK:
T2763-100G:
T2763-25G:4548173996844
T2763-5G:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service