Skip to Content
Merck
All Photos(1)

Key Documents

SML1831

Sigma-Aldrich

KYA1797K potassium salt

≥98% (HPLC)

Synonym(s):

(5Z)-5-[[5-(4-Nitrophenyl)-2-furanyl]methylene]-4-oxo-2-thioxo-3-thiazolidinepropanoic acid potassium salt, (Z)-3-(5-((5-(4-nitrophenyl)furan-2-yl)methylene)-4-oxo-2-thioxothiazolidin-3-yl)propanoic acid potassium salt, KYA1797 potassium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H11KN2O6S2
CAS Number:
Molecular Weight:
442.51
UNSPSC Code:
12352200

Assay

≥98% (HPLC)

form

powder

color

orange to dark red

solubility

DMSO: 2 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

[O-]C(CCN1C(/C(SC1=S)=C/C2=CC=C(C3=CC=C([N+]([O-])=O)C=C3)O2)=O)=O.[K+]

Biochem/physiol Actions

Inhibitor of Wnt/β-catenin and Ras pathways by enhancing formation of the β-catenin destruction complex
KYA1797K is an inhibitor of both Wnt/β-catenin and Ras pathways. It binds to the axin regulators of the G-protein signaling (RGS) domain enhancing formation of the β-catenin destruction complex and inducing activation GSK3β, which results in phosphorylation and subsequent degradation of both β-catenin and K-Ras. KYA1797K was show to suppress tumor growth and progression in mouse xenografts of CRC cells that had APC and K-Ras mutations.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pu-Hyeon Cha et al.
Nature chemical biology, 12(8), 593-600 (2016-06-14)
Both the Wnt/β-catenin and Ras pathways are aberrantly activated in most human colorectal cancers (CRCs) and interact cooperatively in tumor promotion. Inhibition of these signaling may therefore be an ideal strategy for treating CRC. We identified KY1220, a compound that
Robert Lodge et al.
mBio, 14(5), e0195023-e0195023 (2023-09-29)
In order to efficiently produce infectious viral particles, HIV must counter several restrictions exerted by host cell antiviral proteins. MARCH1 is a member of the MARCH protein family that restricts HIV infection by limiting the incorporation of viral envelope glycoproteins

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service