Skip to Content
Merck
All Photos(1)

Documents

Safety Information

P3895

Sigma-Aldrich

Phosphatase, Alkaline from human placenta

≥10 units/mg solid (in glycine buffer)

Synonym(s):

Orthophosphoric-monoester phosphohydrolase (alkaline optimum)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

form

solid

specific activity

≥10 units/mg solid (in glycine buffer)

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... ALPP(250)

Looking for similar products? Visit Product Comparison Guide

Application

Alkaline phosphatase is used for conjugation to antibodies and other proteins for ELISA, Western blotting, and histochemical detection. It is routinely used to dephosphorylate proteins and nucleic acids. It may be used for protein labeling when high sensitivity is required. Alkaline phosphatase may be also be used to dephosphorylate the 5′-termini of DNA or RNA to prevent self-ligation. DNA or RNA can also be tagged with radiolabeled phosphate (via T4 polynucleotide kinase) after dephosphorylation with alkaline phosphatase.

Biochem/physiol Actions

Alkaline phosphatase dephosphorylates proteins and nucleic acids. Human placental alkaline phosphatase (PLAP) has an unique residue, Glu-429. It is an allosteric enzyme that is uncompetitively inhibited by L-Phe, L-Trp, and L-Leu. PLAP is post-translationally modified. Phosphatidylinositol-specific phospholipase C may be involved in the release of PLAP. PLAP may have a role in cell division and may be involved in the transfer of maternal IgG to the fetus.

Unit Definition

One unit will hydrolyze 1 μmole of 4-nitrophenyl phosphate per minute at pH 10.4 at 37 °C.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

P3895-BULK:
P3895-100MG:
P3895-10MG:
P3895-VAR:
P3895-25MG:
P3895-250MG:
P3895-5MG:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yongliang Yang et al.
Chemical biology & drug design, 78(6), 923-931 (2011-09-14)
Human placental alkaline phosphatase has been identified as a hydrolase that is significantly overexpressed on the surface of various solid tumor cells, and is therefore a suitable prodrug design target for non-invasive cancer imaging and therapy. Structure-based prediction of enzymatic
Mary F O'Leary et al.
Nutrients, 13(12) (2021-12-29)
Shatavari has long been used as an Ayurvedic herb for women's health, but empirical evidence for its effectiveness has been lacking. Shatavari contains phytoestrogenic compounds that bind to the estradiol receptor. Postmenopausal estradiol deficiency contributes to sarcopenia and osteoporosis. In
M H Le Du et al.
The Journal of biological chemistry, 276(12), 9158-9165 (2001-01-01)
Human placental alkaline phosphatase (PLAP) is one of three tissue-specific human APs extensively studied because of its ectopic expression in tumors. The crystal structure, determined at 1.8-A resolution, reveals that during evolution, only the overall features of the enzyme have
Yixing Cheng et al.
Journal of biomedical nanotechnology, 10(2), 287-298 (2014-04-18)
Scaffold architecture, surface topography, biochemical and mechanical cues have been shown to significantly improve cellular events and in vivo tissue regeneration. Specifically electrospun nanofiber matrices have gained tremendous interest due to their intrinsic structural resemblance to native tissue extracellular matrix
Jean-Paul Lallès
Nutrition reviews, 72(2), 82-94 (2014-02-11)
Important protective roles of intestinal alkaline phosphatase (IAP)--including regulation of intestinal surface pH, absorption of lipids, detoxification of free nucleotides and bacterial lipopolysaccharide, attenuation of intestinal inflammation, and possible modulation of the gut microbiota--have been reviewed recently. IAP is modulated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service