Skip to Content
Merck
All Photos(1)

Documents

Safety Information

M6324

Sigma-Aldrich

MART-1 (27-35), Human

Synonym(s):

Antigen LB39-AA (27-35) (human), Antigen SK29-AA (27-35) (human), Melan-A Protein (27-35) (human), Melanoma Antigen Recognized By T-Cells 1 (27-35) (human)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

form

lyophilized powder

Quality Level

color

white

solubility

water, double-distilled: soluble

storage temp.

−20°C

Amino Acid Sequence

Ala-Ala-Gly-Ile-Gly-Ile-Leu-Thr-Val

General description

The peptide represents amino acids 27-35 of MART-1 protein that is the melanoma antigen recognized by T cells. This peptide mimics the natural epitope that is presented by T cells in humans with reactivity to melanoma cells.
Melanoma-associated antigen recognized by T cells-1 (MART-1), also known as Melan-A, is encoded by the gene mapped to human chromosome 9p24.1. The encoded protein is a hydrophobic transmembrane protein. MART-1 is mainly expressed in skin and retinae melanocytes.

Biochem/physiol Actions

Melanoma-associated antigen recognized by T cells-1 (MART-1) is essential for the functioning of glycoprotein-100 (gp100). Overexpression of the gene has been observed in cutaneous melanoma (CM). In addition, deletion of Mart-1 gene leads to parotid metastatic melanoma.

Physical form

Lyophilized from 0.1% aqueous TFA

Other Notes

Lyophilized from 0.1% TFA in H2O

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

M6324-VAR:
M6324-1MG:
M6324-BULK:
M6324-5MG-PW:
M6324-1MG-PW:
M6324-5MG:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Comparative genomic hybridization in a case of melanoma that loses expression of S100, HMB45, Melan A and tyrosinase in metastasis
Guo R, et al.
International Journal of Clinical and Experimental Pathology, 7(1), 468-468 (2014)
María Marcela Barrio et al.
PloS one, 7(7), e40311-e40311 (2012-07-07)
Dendritic cells (DC) can achieve cross-presentation of naturally-occurring tumor-associated antigens after phagocytosis and processing of dying tumor cells. They have been used in different clinical settings to vaccinate cancer patients. We have previously used gamma-irradiated MART-1 expressing melanoma cells as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service