Skip to Content
Merck
All Photos(5)

Key Documents

Safety Information

CWDS-APC

Sigma-Aldrich

ColorWheel® Antibody-Ready Allophycocyanin (APC) Dye

greener alternative

for use with ColorWheel® Dye-ready Antibodies (Required, sold separately)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116015
NACRES:
NA.41
Product CWDS-APC is not currently sold in your country. Contact Technical Service

product line

ColorWheel®

Quality Level

form

lyophilized

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

technique(s)

flow cytometry: suitable

fluorescence

λex 651 nm; λem 660 nm

compatibility

for use with ColorWheel® Dye-ready Antibodies (Required, sold separately)

greener alternative category

shipped in

ambient

storage temp.

2-8°C

General description

ColorWheel® technology is a novel and proprietary method of creating your own antibody and dye combinations for use in flow cytometry. By incubating any ColorWheel® antibody with any ColorWheel® dye, researchers can quickly and simply produce primary conjugated antibodies for use in single color or multicolor flow cytometry analysis. Each ColorWheel® antibody and dye is lyophilized for long long-term storage, allowing for simplicity and flexibility without compromise. ColorWheel® technology requires both ColorWheel® antibody and ColorWheel® dye for Flow Cytometry application.

Allophycocyanin (APC) belongs to a family of phycobiliproteins and is well studied as a fluorescent reagent for flow cytometric analysis. It is isolated from a Spirulina species, a type of blue-green alga and is composed of α and β subunits. The heterodimers of these subunits can assemble as trimer or hexamer. APC displays high absorptivity and quantum efficiency. It has an excitation max at 651 nm and a emission max at 660 nm with extinction coefficient of 700,000 cm-1 M-1. It can absorb and emit light with low energy losses to quenching. APC can be easily conjugated to monoclonal and polyclonal antibodies for use in fluorescence-based detection methods, such as flow cytometry and FACS analysis. (Ref.: Sonani, RR., et al. (2015). PLoS One. 10(4): e0124580; Jung, TM., and Daily MO. (1989). J. Immunol. Methods. 121(1); 9-18).
We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This product is Preservative-free, lyophilized product for enhanced stability and allow for ambient shipping and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.

Application

Flow Cytometry: Excitation: 651 nm, Emission: 660 nm

Compatibility

Requires ColorWheel® Antibody (Sold Separately)

Physical form

CWDS-APC-25UL Set Components:

CWD-APC-25UL: ColorWheel® Antibody-Ready Allophycocyanin (APC) Dye
Lyophilized from PBS containing D-Mannitol and Sucrose, normal appearance is a dried pellet. Reconstituted solution is stable and functional as assessed by functional testing. Contains no biocide or preservatives, such as azide.

CWD-FINISH-25UL: ColorWheel® Finishing Buffer
Lyophilized from PBS. Contains no biocide or preservatives, such as azide.

Reconstitution

Reconstitute each vial by adding 25 μL PBS. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C. Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial. Reconstitute each vial by adding 25 μL of PBS. Protect from light.

Legal Information

ColorWheel is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

11 - Combustible Solids


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

PDSCL

Please refer to KIT Component information

PRTR

Please refer to KIT Component information

FSL

Please refer to KIT Component information

ISHL Indicated Name

Please refer to KIT Component information

ISHL Notified Names

Please refer to KIT Component information

Cartagena Act

Please refer to KIT Component information

JAN Code

キットコンポーネントの情報を参照してください


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service