Skip to Content
Merck
All Photos(1)

Documents

Safety Information

AV36360

Sigma-Aldrich

Anti-RECQL5 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-FLJ90603, Anti-RECQ5, Anti-RecQ protein-like 5

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

109 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RECQL5(9400)

Immunogen

Synthetic peptide directed towards the middle region of human RECQL5

Biochem/physiol Actions

RECQL5 is an important backup helicase that maintains the genomic stability. It displaces RAD51 from ssDNA and prevents aberrant homologous recombination. The expression of RECQL5 is significantly decreased in sporadic primary colorectal cancers and acts as a marker to identify these cancers that are susceptible to chemotherapy.

Sequence

Synthetic peptide located within the following region: CDHCQNPTAVRRRLEALERSSSWSKTCIGPSQGNGFDPELYEGGRKGYGD

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

AV36360-50UG:
AV36360-100UL:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Venkateswarlu Popuri et al.
Critical reviews in biochemistry and molecular biology, 48(3), 289-299 (2013-05-01)
DNA helicases are ubiquitous enzymes that catalyze unwinding of duplex DNA and function in all metabolic processes in which access to single-stranded DNA is required, including DNA replication, repair, recombination and RNA transcription. RecQ helicases are a conserved family of
Victoria Valinluck Lao et al.
Translational oncology, 6(4), 458-469 (2013-08-03)
Deregulation of DNA repair enzymes occurs in cancers and may create a susceptibility to chemotherapy. Expression levels of DNA repair enzymes have been shown to predict the responsiveness of cancers to certain chemotherapeutic agents. The RECQ helicases repair damaged DNA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service