This kit is specific for Capsase 3 (CASP-3), a member of the Caspase family (cysteine-aspartic acid protease) and when activated, is an essential factor in cell apoptosis. The Cut-N-Glow approach involves the introduction of a structural distortion into one of the complementary fragments (GFP 11), through the use of a conditionally stable tether, which serves to constrain the N and C termini of GFP 11 closely in space, thereby, diminishing the mutual affinity of the two fragments and blocking protein self-assembly until the tether is cleaved. The distortion can be reversed upon site-specific proteolysis of the tether, resulting in GFP 11 fragment assembly with GFP 1-10, generating reconstituted, functional GFP. Depending on the tether, the chimeric GFP can serve as a substrate for proteases from the three major classes: serine, cysteine, and aspartic acid.
Suitability
This kit contains sufficient reagents for one 96 well plate (96 tests)
Storage Class Code
10 - Combustible liquids
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
Laboratory investigation; a journal of technical methods and pathology, 102(5), 515-523 (2022-01-20)
Mitochondrial dynamics and quality control play a central role in the maintenance of the proliferation-apoptosis balance, which is closely related to the progression of pulmonary arterial hypertension (PAH). However, the exact mechanism of this balance remains unknown. Pulmonary artery smooth
Cellular apoptosis assays to detect programmed cell death using Annexin V, Caspase and TUNEL DNA fragmentation assays.
Questions
Reviews
★★★★★ No rating value
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.