Skip to Content
Merck
All Photos(2)

Documents

Safety Information

223697

Sigma-Aldrich

Potassium hydrogensulfate, fused

ACS reagent, Mixture of K2S2O7 and KHSO4

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NB.24

grade

ACS reagent

Quality Level

form

powder

concentration

37.5-38.6% H2SO4 in NaOH (titration)

impurities

≤0.002% H2SO4
≤0.01% insolubles
≤2.5% water

acid number

37.5‑38.6%

density

2.512 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤0.002%
phosphate (PO43-): ≤0.001%

cation traces

Ca: ≤0.002%
Fe: ≤0.002%
Mg: ≤0.001%
Na: ≤0.01%
heavy metals: ≤0.001%

SMILES string

[K+].[K+].[K+].OS([O-])(=O)=O.[O-]S(=O)(=O)OS([O-])(=O)=O

InChI

1S/3K.H2O7S2.H2O4S/c;;;1-8(2,3)7-9(4,5)6;1-5(2,3)4/h;;;(H,1,2,3)(H,4,5,6);(H2,1,2,3,4)/q3*+1;;/p-3

InChI key

ZXJZSJHWYNTTSK-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Eye Dam. 1 - Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

ISHL Indicated Name

Substances Subject to be Indicated Names

ISHL Notified Names

Substances Subject to be Notified Names

JAN Code

223697-VAR:
223697-1KG:
223697-5G:
223697-250G:
223697-BULK:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Electrochemical Behavior of Molten V2O5- K2S207-KHSO4 Systems.
Petrushina IM, et al.
Journal of the Electrochemical Society, 144(2), 532-539 (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service