75349
Oligo dT-Cellulose
suitable for affinity chromatography
Synonym(s):
Oligothymidylic acid, immobilized on cellulose
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
MDL number:
UNSPSC Code:
12190000
Recommended Products
technique(s)
affinity chromatography: suitable
capacity
~20 A260 units/g, material capacity (poly A)(pH 7.5 in 0.5 M NaCl, 0°C)
storage temp.
−20°C
General description
mixture of oligothymidylic acids (up to 18 nucleotides) covalently attached to cellulose via the terminal 5′-phosphoryl end by the method of P.T. Gilham, JACS 86, 4982 (1964)
Other Notes
Isolation of polyadenylated RNA; Purification of mRNA; Purifying RNA by column chromatography
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Purifying RNA by column chromatography.
N K Tanner
Methods in enzymology, 180, 25-41 (1989-01-01)
Effect of flow rate on the isolation of polyadenylated RNA on oligo(dT)-cellulose columns.
S Nadin-Davis et al.
Journal of biochemical and biophysical methods, 11(2-3), 185-189 (1985-08-01)
Nikolay E Shirokikh et al.
Proceedings of the National Academy of Sciences of the United States of America, 105(31), 10738-10743 (2008-07-29)
Eukaryotic mRNAs in which a poly(A) sequence precedes the initiation codon are known to exhibit a significantly enhanced cap-independent translation, both in vivo and in cell-free translation systems. Consistent with high expression levels of poxviral mRNAs, they contain poly(A) sequences
High-efficiency cloning of full-length cDNA; construction and screening of cDNA expression libraries for mammalian cells.
H Okayama et al.
Methods in enzymology, 154, 3-28 (1987-01-01)
H Aviv et al.
Proceedings of the National Academy of Sciences of the United States of America, 69(6), 1408-1412 (1972-06-01)
A convenient technique for the partial purification of large quantities of functional, poly(adenylic acid)-rich mRNA is described. The method depends upon annealing poly(adenylic acid)-rich mRNA to oligothymidylic acid-cellulose columns and its elution with buffers of low ionic strength. Biologically active
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service