Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

68698

Supelco

Myristic-d27 acid

analytical standard

Synonym(s):

Tetradecanoic-d27 acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CD3(CD2)12CO2H
CAS Number:
Molecular Weight:
255.54
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

isotopic purity

≥98 atom % D

Assay

≥99.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

250 °C/100 mmHg (lit.)

mp

52-54 °C (lit.)
53-55 °C

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

functional group

carboxylic acid

mass shift

M+27

shipped in

ambient

storage temp.

room temp

SMILES string

[2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C(O)=O

InChI

1S/C14H28O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14(15)16/h2-13H2,1H3,(H,15,16)/i1D3,2D2,3D2,4D2,5D2,6D2,7D2,8D2,9D2,10D2,11D2,12D2,13D2

InChI key

TUNFSRHWOTWDNC-RZVOLPTOSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

68698-25MG:
68698-BULK:
68698-VAR:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nathan A Palmer et al.
PloS one, 9(8), e105138-e105138 (2014-08-19)
Switchgrass (Panicum virgatum L.) is being developed as a bioenergy crop for many temperate regions of the world. One way to increase biomass yields is to move southern adapted lowland cultivars to more northern latitudes. However, many southerly adapted switchgrass
Ning Chen et al.
Cytotechnology, 66(6), 945-966 (2013-12-03)
Optimization of cell culture processes can benefit from the systematic analysis of experimental data and their organization in mathematical models, which can be used to decipher the effect of individual process variables on multiple outputs of interest. Towards this goal
Zhihong Niu et al.
The Journal of clinical endocrinology and metabolism, 99(11), E2269-E2276 (2014-04-04)
Both polycystic ovary syndrome (PCOS) and obesity are associated with specific reproductive health complications, including lower oocyte quality and clinical pregnancy rates in assisted conception cycles, which may be a result of metabolism-induced changes in the oocyte through the microenvironment
Syed Ghulam Musharraf et al.
Scientific reports, 5, 8607-8607 (2015-02-26)
Lung cancer has been the most common death causing cancer in the world for several decades. This study is focused on the metabolite profiling of plasma from lung cancer (LC) patients with three control groups including healthy non-smoker (NS), smokers
M Arbulu et al.
Analytica chimica acta, 858, 32-41 (2015-01-20)
The current study presents a method for comprehensive untargeted metabolomic fingerprinting of the non-volatile profile of the Graciano Vitis vinifera wine variety, using liquid chromatography/electrospray ionization time of flight mass spectrometry (LC-ESI-QTOF). Pre-treatment of samples, chromatographic columns, mobile phases, elution

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service