Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

05-5560

Sigma-Aldrich

Colour standard solution

JIS (color standard), JIS: 1000

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
¥5,770

¥5,770


In StockDetails


Request a Bulk Order

Select a Size

Change View
100 ML
¥5,770

About This Item

UNSPSC Code:
41116107

¥5,770


In StockDetails


Request a Bulk Order

Agency

JIS (color standard)

availability

available only in Japan

color

JIS: 1000

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Carc. 1B - Met. Corr. 1

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

ISHL Indicated Name

Substances Subject to be Indicated Names

ISHL Notified Names

Substances Subject to be Notified Names

JAN Code

05-5560-6-100ML-J:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service