Recommended Products
material
polysulfone flask (receiver)
sterility
sterile
manufacturer/tradename
Sterifil®
parameter
250 mL sample volume
technique(s)
vacuum filtration: suitable
diam.
47 mm
application(s)
filtration
compatibility
for use with Sterifil Aseptic System
shipped in
ambient
General description
The Sterifil® aseptic system was designed as a tool for general filtration applications and filtration of samples for particulate or biological contamination analysis. The closed unit protects sample and filtrate from environmental contamination. Sterifil holder and funnel are available separately (without receiver flask and cover) to use with standard 1 L filter flasks or multiple place manifolds.
Depending upon membrane filter selection, the Sterifil products can be used for a variety of applications including bioburden monitoring and enumeration of organisms in samples.
Features & Benefits:
•Closed unit protects samples and filtrate from environmental contamination
•Device is autoclavable with membrane in place for sterile applications with less waste
•Optional hand pump allows for on-site filtration
Applications:
General Filtration, Field-based Filtration (with optional hand pump), Bioburden Monitoring, Counting Microorganisms, Filtration Processes Requiring Membrane to be Retained
Depending upon membrane filter selection, the Sterifil products can be used for a variety of applications including bioburden monitoring and enumeration of organisms in samples.
Features & Benefits:
•Closed unit protects samples and filtrate from environmental contamination
•Device is autoclavable with membrane in place for sterile applications with less waste
•Optional hand pump allows for on-site filtration
Applications:
General Filtration, Field-based Filtration (with optional hand pump), Bioburden Monitoring, Counting Microorganisms, Filtration Processes Requiring Membrane to be Retained
Legal Information
Sterifil is a registered trademark of Merck KGaA, Darmstadt, Germany
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service