Skip to Content
Merck
All Photos(1)

Key Documents

NSCAP50

Millipore

NovaSeptum® GO Conical Tube 50mL Centrifugation Cap

sterile; γ-irradiated

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104105

Agency

according to ISO 11137 (sterilization)
according to ISO Clean Room Class 8 conditions in a facility
certified by the ISO 14644-1 standards
meets USP 88 biological reactivity (all component materials)

Quality Level

sterility

sterile; γ-irradiated

product line

NovaSeptum® GO

feature

autoclavable: no

parameter

max. pressure at 0.50 bar (7.25 psi)
2-60 °C temp. range (36-140 °F)

application(s)

cell therapy
gene therapy
life science and biopharma
mAb
ophthalmics
parenterals
pharma/biopharma processes
sterile sampling
viral therapy

storage temp.

2-30°C

General description

Device configuration: single Unit Sampling
NovaSeptum® GO Conical Tubes are small volume sampling containers that fit our established (TM="NovaSeptum"] GO sampling system. These sampling containers are available in two sizes: 15 mL polystyrene tubes ideal for endotoxin testing, and 50 mL polypropylene tubes for bioburden and other physico-chemical tests. These conical tubes provide flexibility to manufacturers for sampling at any point in the production process eliminating the need to aliquot.

NovaSeptum® GO Conical Tubes are available in both standard and custom options. When used with the optional accessory sterile cap, NovaSeptum® GO Conical Tubes can be centrifuged directly, eliminating the need for sample transfer.

The NovaSeptum® GO Conical Tube Sampling Unit offers you flexibility while minimizing the risk of both product and process contamination.

Application

Sterility testing, Bioburden, pyrogenic testing, endotoxin testing, chemical analysis, pH analysis, fermentation applications or any other routine lab testing
supported liquid extraction

Features and Benefits

Sterile cap for centrifugation

Packaging

10 caps per box

Components

HDPE (High Density Polyethylene)

Legal Information

NOVASEPTUM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

NovaSeptum® GO sterile sampling unit with 15 mL single-use conical tube for small volume and endotoxin sampling. Closed design for aseptic process. Reduce product loss and risks of contamination. Available in single and manifold sampling format. Easily integrated into stainless-steel system via multi-use connection types (TC, in-line, ingold®).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service